D-(-)-Ornithine hydrochloride ~99%, white crystalline powder

Supplier: MP Biomedicals

Synonyms: H-D-Orn-OH·HCl, H-D-Orn-OH.HCl

0210251401 0210251405 0210251410
ICNA0210251401EA 41.4 EUR
ICNA0210251401 ICNA0210251405 ICNA0210251410
D-(-)-Ornithine hydrochloride ~99%, white crystalline powder
D(-)-Ornithine hydrochloride

D-Ornithine cleave proteins at cysteine residues.


Soluble in water (30 mg/ml - clear, colorless solution).


Caution: Research Use Only (RUO).

Formula: C₅H₁₂N₂O₂·HCl
MW: 168,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012917
CAS Number: 16682-12-5

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 235 - 245 °C
Optical Rotation Report Result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR