Uric acid ∼99%, white powder

Supplier: MP Biomedicals

Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione

0210321510 0210321580 0210321590
ICNA0210321510EA 38.9 EUR
ICNA0210321510 ICNA0210321580 ICNA0210321590
Uric acid ∼99%, white powder
Uric acid

Soluble in alkaline solutions (1% in 1 N NaOH), glyurol; very slightly to insoluble in water; insoluble in ethanol, ether and alcohol.


Soluble in alkaline solutions (1% in 1 N NaOH), glyurol; very slightly to insoluble in water; insoluble in ethanol, ether and alcohol.


Caution: Research Use Only (RUO).

Formula: C₅H₄N₄O₃
MW: 168.11 g/mol
Melting Pt: 436
Storage Temperature: Ambient
MDL Number: MFCD00005712
CAS Number: 69-93-2
Merck Index: 12,10014

Order Now

Specification Test Results

Purity ~99%
FTIR Conforms to Standard
Loss on Drying ≤0,5%
UV/VIS: λmax (water) 230-235, 285-290 nm
Residue on Ignition <0,2%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR