3-O-Methyl-α-D-glucopyranose, white powder
Supplier: MP Biomedicals
Synonyms:
3-o-Methyl-D-glucose
Marker to assess glucose transport.
Varying concentrations of 3-O-methyl-D-glucopyranose were used to measure responses of jejunal and ileal short circuit currents to determine maximal transport capacity and carrier affinity in a study to assess intestinal glucose transport in rats with diabetes mellitus. It has also been used in a study to investigate nonmetabolizable glucose analogues and ornithine decarboxylase expression in LLC-PK1 cells.
Caution: Research Use Only (RUO).
Formula:
C₇H₁₄O₆ Storage Temperature: Ambient |
CAS Number:
13224-94-7 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 160 - 170 °C |
Optical Rotation | +53° to +61° (c=1, water, ammonium hydroxide) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...