Fosfomycin disodium salt

Supplier: MP Biomedicals

Synonyms: Disodium (1R,2S)-1,2-Epoxypropylphosphonate, Phosphomycin Disodium Salt, (1R,2S)-1,2-Epoxypropylphosphonic Acid Disodium Salt

0215187601 0215187605 0215187610
ICNA0215187601EA 46.9 EUR
ICNA0215187601 ICNA0215187605 ICNA0215187610
Fosfomycin disodium salt
Fosfomycin disodium salt

Soluble in water (50 mg/ml) clear,colourless solution.


  • Soluble in water (50 mg/ml)
  • Clear, colourless solution

Formula: C₃H₅Na₂O₄P
MW: 182.02 g/mol
MDL Number: MFCD00056853
CAS Number: 26016-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR