Methyl-ɑ-D-mannopyranoside ∼99%, white crystalline powder
Supplier: MP Biomedicals
Synonyms:
ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
Used to reduce Con A binding to lymphocyte surface in culture.
Methyl α-D-mannopyranoside has been used to synthesize a series of tri- and tetrahydroxylated seven-membered iminosugars in a study that worked towards a stable noeuromycin analog with a D-manno configuration. It has also been used in a study to investigate the primary mannose binding site of pradimicin A.
Soluble in water.
Caution: Research Use Only (RUO).
Formula:
C₇H₁₄O₆ MW: 194,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063262 CAS Number: 617-04-9 |
Specification Test Results
Identity Test | Passes |
Purity | ~99% |
FTIR | Conforms to Standard |
Melting Point | 187 - 197 °C |
Loss on Drying | ≤0.5% |
Optical Rotation | +76 to +85 °C (c=1, water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...