1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride, white powder
Supplier: MP Biomedicals
Synonyms:
Meclizine dihydrochloride
0215534105
0215534125
ICNA0215534105EA
54.1
EUR
ICNA0215534105
ICNA0215534125
1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride, white powder
1-((4-Chlorophenyl)(phenyl)methyl)-4-(3-methylbenzyl)piperazine dihydrochloride
Formula:
C₂₅H₂₉Cl₃N₂ MW: 463,87 g/mol |
MDL Number:
MFCD00058199 CAS Number: 1104-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...