13-cis-Retinoic acid, orange powder

Supplier: MP Biomedicals

Synonyms: Isotretinoin, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid

0215651880 0215651883 0215651890
ICNA0215651880EA 113 EUR
ICNA0215651880 ICNA0215651883 ICNA0215651890
13-cis-Retinoic acid, orange powder
13-cis-Retinoic acid

13-cis-Retinoic acid (RA) has anti-inflammatory and anti-tumor action.


The action of 13-cis-Retinoic acid (RA) is mediated through RAR-β and RAR-α receptors. RA attenuates iNOS expression and activity in cytokine-stimulated murine mesangial cells. It induces mitochondrial membrane permeability transition, observed as swelling and as a decrease in membrane potential, and stimulates the release of cytochrome c implicating mechanisms through the apoptosis pathway. These activities are reversed by EGTA and cyclosporin A. RA also increases MMP-1 protein expression partially via increased transcription.


Soluble in chloroform (50 mg/ml); sparingly soluble in ether; slightly soluble in ethanol and isopropanol; practically insoluble in water.

Formula: C₂₀H₂₈O₂
MW: 300,44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00079542
CAS Number: 4759-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR