Lincomycin hydrochloride ≥82%, powder

Supplier: MP Biomedicals

0215894801 0215894805
ICNA0215894801EA 85.3 EUR
ICNA0215894801 ICNA0215894805
Lincomycin hydrochloride ≥82%, powder
Lincomycin hydrochloride

Lincomycin hydrochloride is used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding.


Lincomycin is a lincosamide antibiotic that forms cross-links within the peptidyl transferase loop region of the 23S rRNA. Mode of action: inhibits bacterial protein synthesis; antimicrobial spectrum: gram-positive bacteria.


Soluble in water, ethanol, methanol, DMSO (25 mg/ml).

Formula: C₁₈H₃₄N₂O₆S·HCl
MW: 443.0 g/mol (anhy)
MDL Number: MFCD00058237
CAS Number: 859-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR