Clozapine, yellow powder

Supplier: MP Biomedicals

Synonyms: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine

0215976005 0215976050
ICNA0215976005EA 62.6 EUR
ICNA0215976005 ICNA0215976050
Clozapine, yellow powder
Clozapine

Clozapine is a dibenzodiazepine compound, structurally related to loxapine is used in cell signaling research. It is an a typical antipsychotic medication used in the treatment of schizophrenia. It is a dopamine D4 and D2 receptor antagonist which has high affinity for the cloned rat dopamine D4 receptor (Ki < 20 nM). It is also a potent, selective muscarinic antagonist which exhibits the highest reported affinity for 5-HT1A and 5-HT1C sites.


Clozapine has dopamine receptor blocking activity, α-adrenergic blocking, antimuscarinic, antihistamine, and antiserotonergic properties. The role of clozapine in reversing the inhibitory effect of dopamine on sodium and water transport in the rat cortical collecting duct has been reviewed. Clozapine has been used to probe the role of the calcium/calmodulin-dependent kinase II in NMDA-induced inward currents and electrically evoked excitatory postsynaptic currents in mouse brain sections, cultured mouse cells, and transgenic mice.


Soluble in 0,1 M HC1 (25 mg/ml) or DMSO (100 mM).

Formula: C₁₈H₁₉ClN₄
MW: 326,83 g/mol
Melting Pt: 182…184 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00153785
CAS Number: 5786-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR