Estradiol benzoate, powder

Supplier: MP Biomedicals
Danger

Synonyms: β-Estradiol-3-benzoate

0219041101 0219041105
ICNA0219041101EA 78.65 EUR
ICNA0219041101 ICNA0219041105
Estradiol benzoate, powder
Estradiol benzoate

Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.


Soluble in alcohol, acetone, dioxane, chloroform(50 mg/ml); slightly soluble in ether, vegetable oils.

Formula: C₂₅H₂₈O₃
MW: 376,496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003692
CAS Number: 50-50-0

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 190 - 200°C
Optical Rotation (c=1, CHCl3) +60 ± 3°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR