Microbe creatinine deiminase, MP Biomedicals

Supplier: MP Biomedicals

0215349025 0215349080 0215349083 0215349091
ICNA0215349025EA 156 EUR
ICNA0215349025 ICNA0215349080 ICNA0215349083 ICNA0215349091
Microbe creatinine deiminase, MP Biomedicals
Enzymes

Creatinine deiminase has been used in a study to assess the application of a creatinine-sensitive biosensor for hemodialysis control. It has also been used in a study to investigate the bioelectronic tongue for the simultaneous determination of urea, creatinine and alkaline ions in clinical samples.


Enzyme Commission Number: E.C. 3.5.4.21
Isoelectric point (pI): 4,4 (Lit.)
Michaelis constant : 3,5×10−3 M (Creatinine)
Thermal stability : below 65 °C (pH 7,5)
Optimum temperature : 65 to 75 °C


Unit definition: One unit causes the formation of one µmole of ammonia (the oxidation of one µmole of NADPH) per minute at pH 7,5 at 37 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR