myo-Inositol

Supplier: MP Biomedicals

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

0210205250 0210205280 0210205290 0210205291
ICNA0210205250EA 53 EUR
ICNA0210205250 ICNA0210205280 ICNA0210205290 ICNA0210205291
myo-Inositol
myo-Inositol

D-myo-INOSITOL (i-Inositol; myo-Inositol; meso-Inositol) A lipotropic agent.


pH: Neutral to Litmus (10% aq soln)

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 291
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
Merck Index: 13,05001

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR