Anti-EC SOD Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
ADISOD106D ADISOD106F
ENZOADISOD106DEA 241 EUR
ENZOADISOD106D ENZOADISOD106F
Anti-EC SOD Rabbit Polyclonal Antibody
Antibodies
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

Type: Primary
Antigen: EC SOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
New Product
Order Now

SPECIFICATIONS

Antigen symbol EC SOD
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes
Immunogen Synthetic peptide corresponding to a portion of EC SOD. The sequence is completely conserved in human, mouse, rat and rabbit.
Form Liquid
Purification Purified by Protein A affinity chromatography
Storage buffer PBS containing 50% glycerol and 0,09% sodium azide
Storage temperature −20 °C
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR