Human recombinant Annexin V, FITC (Fluorescein Isothiocyanate)
Supplier: Enzo Life Sciences
ALX2092563100 ALX209256T100
ENZOALX2092563100EA 681 EUR
ENZOALX2092563100 ENZOALX209256T100
Human recombinant Annexin V, FITC (Fluorescein Isothiocyanate)
Proteins and Peptides
FITC-labelled recombinant human annexin V fusion protein (produced in E. coli) shows bright green fluorescence (Ex(max): 488 nm; Em(max): 530 nm).
Ordering information: Quantity: Sufficient for 100 or 300 tests. 1 test is equivalent to 1μg of the conjugated protein in an experimental volume of 0.4 ml.