Mouse nerve growth factor 25S (from submaxillary glands)
Supplier: ENZO LIFE SCIENCES
ALX521007C025
ALX521007C100
ENZOALX521007C025EA
330
EUR
ENZOALX521007C025
ENZOALX521007C100
Mouse nerve growth factor 25S (from submaxillary glands)
Proteins and Peptides
Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984).
Ordering information: Quantity: Concentration is estimated spectroscopically from its extinction coefficient and amino acid analysis.