3-Methylxanthine, white powder
Supplier: MP Biomedicals
|
Warning
|
3-Methylxanthine, a metabolite of theophylline, was quantitated in rat plasma by a sensitive LC-MS/MS method.
Freely soluble in alkali hydroxides and in mineral acid; soluble in dilute aqueous base or ethanol; slightly soluble in hot water; practically insoluble in organic solvent
Formula:
C₆H₆N₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00005580 CAS Number: 1076-22-8 EINECS: 214-058-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...