PIPES sesquisodium salt
Supplier: MP Biomedicals
Synonyms:
2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid sesquisodium salt, Sesquisodium piperazine-1,4-diethanesulphonate, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulphonate 1-(4-(1-sulphoethyl)piperazin-1-yl)ethanesulphonate(3:2), PIPES-Na1,5, Piperazine-N,N'-bis-2-ethanesulphonic acid sesquisodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) sesquisodium salt
PIPES free acid is not very water soluble (approximately 1 g/L at 100 °C) but will solubilise in 0,5 M NaOH.
Effective buffering range : 6,1 - 7,5; ΔpK/ΔT: 0,0085(Lit.)
Formula:
C₁₆H₃₃N₄Na₃O₁₂S₄ Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066393 CAS Number: 100037-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...