U-69593 (synthetic) ≥99% (by TLC), white solid
Supplier: ENZO LIFE SCIENCES
BMLN7050005
BMLN7050025
ENZOBMLN7050005EA
165
EUR
ENZOBMLN7050005
ENZOBMLN7050025
U-69593 (synthetic) ≥99% (by TLC), white solid
U-69593
Opioid receptor ligand.
Selective κ-opioid agonist. Thought to be selective for the κ-1 subtype (KD=3.8 nM in human cerebral cortex membranes). Immunomodulatory activity.
Soluble in 100% ethanol, 0.1 N hydrogen chloride, or DMSO. Insoluble in water or 0.1 N sodium hydroxide.
Formula:
C₂₂H₃₂N₂O₂ MW: 356.51 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD05664586 CAS Number: 96744-75-1 |
Specification Test Results
Purity | ≥99% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...