7,7-Dimethyleicosadienoic acid ≥99%

Supplier: ENZO LIFE SCIENCES

BMLEI1120010 BMLEI1120050
ENZOBMLEI1120010EA 116 EUR
ENZOBMLEI1120010 ENZOBMLEI1120050
7,7-Dimethyleicosadienoic acid ≥99%
7,7-Dimethyleicosadienoic acid

Phospholipase inhibitor


Non-metabolizable analog of arachidonic or eicosadienoic acid. Cell permeable. DEDA inhibits P388D1 cell phospholipase A2 (PLA2) (IC50=16 µM) and snake venom PLA2 (IC50=14 µM). It inhibits human recombinant sPLA2 (IC50=5.8 µM) selectively over cPLA2. It is also a weak inhibitor of 5-lipoxygenase (IC50=56 µM, RBL-l cells). Inhibits induction of the anti-viral state in human fibroblasts induced by interferon.

Formula: C₂₂H₄₀O₂
MDL Number: MFCD00133010
CAS Number: 89560-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37c21f4b64539d64cb5a3442093ba398af4a393b&vid=860f72af933e7ffda4decb96bf3fc7c233b4917f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR