Geranylgeraniol 95 TLC_ASSAY_METHOD, Oil_Colourless

Supplier: ENZO LIFE SCIENCES
Warning

Synonyms: (2E,6E,10E)-3,7,11,15-Tetramethylhexadeca-2,6,10,14-tetraen-1-ol

BMLG2430020 BMLG2430100
ENZOBMLG2430020EA 114 EUR
ENZOBMLG2430020 ENZOBMLG2430100
Geranylgeraniol 95 TLC_ASSAY_METHOD, Oil_Colourless
Geranylgeraniol

Geranylgeranylpyrophosphate precursor and apoptosis inducer.


Induces apoptosis in HL-60 cells.


Soluble in chloroform or ethyl acetate.

Formula: C₂₀H₃₄O
MW: 290.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00129083
CAS Number: 24034-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbe048b645fdb33fb0def17564b9a83cba1ced3f&vid=6f57638e835f319993613ed0abface1102378b32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (TLC)
Identity Determined by H-NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR