Thioetheramide-phosphorylcholine ≥98% (by TLC)
Supplier: Enzo Life Sciences
A potent substrate analog inhibitor of snake venom PLA2 (IC50=2 µM). It inhibits human recombinant sPLA2 (IC50=0.9 µM) selectively over cPLA2.
- Phospholipase inhibitor
White solid.
Soluble in DMSO, 100% ethanol or methanol.
Formula:
C₄₀H₈₃N₂O₅PS Storage Temperature: Freezer |
CAS Number:
116457-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...