Artemisinin 98

Supplier: ENZO LIFE SCIENCES

Synonyms: Qinghaosu, Arteannuin, (3R,5aS,6R,8aS,9R,12S,12aR)-3,6,9-Trimethyloctahydro-12H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10(3H)-one

ALX350219G001 ALX350219M100
ENZOALX350219G001EA 407 EUR
ENZOALX350219G001 ENZOALX350219M100
Artemisinin 98
Artemisinin

iNOS inhibitor


Powerful antimalarial agent. Inhibits angiogenesis by down-regulating HIF-1α and VEGF expression in mouse embryonic stem cells. Crosses the blood-brain barrier. Inhibitor of human iNOS (NOS II).


Isolated from the traditional Chinese anti-malarial herb Artemisia annua L.

Formula: C₁₅H₂₂O₅
MW: 282.34 g/mol
MDL Number: MFCD00081057
CAS Number: 63968-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR