Azomethine H monosodium salt monohydrate, orange powder

Supplier: MP Biomedicals

0215416905 0215416901 0215416910
ICNA0215416905EA 154 EUR
ICNA0215416905 ICNA0215416901 ICNA0215416910
Azomethine H monosodium salt monohydrate, orange powder
Azomethine H monosodium salt monohydrate

Azomethine H is a reagent for the determination of boron in soil, plants, composts, manure, water and nutrient solutions.


Azomethine H is a reagent for the determination of boron in soil, plants, composts, manure, water and nutrient solutions.


Soluble in hot or cold water (0,1 w/v%-clear).

Formula: C₁₇H₁₂NNaO₈S₂·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149588
CAS Number: 304655-82-1
EINECS: 250-975-3

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR