3-Phenylpropanal ≥95%, clear, colourless liquid
Supplier: MP Biomedicals
|
Warning
|
Synonyms:
3-Phenylpropionaldehyde, Hydrocinnamaldehyde
0215619680
0215619625
ICNA0215619680EA
433
EUR
ICNA0215619680
ICNA0215619625
3-Phenylpropanal ≥95%, clear, colourless liquid
3-Phenylpropanal
Insoluble in water
Formula:
C₉H₁₀O MW: 134,18 g/mol Boiling Pt: 224 °C (1013 hPa) Melting Pt: –42 °C Density: 1,01 g/cm³ (20 °C) |
MDL Number:
MFCD00007021 CAS Number: 104-53-0 EINECS: 203-211-8 |
Specification Test Results
Identity Test | Passes |
Purity | ≥95% |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...