Azure B (azure I), dark green powder, certified C.I. 52010

Supplier: MP Biomedicals

0219052001 0219052010
ICNA0219052001EA 37.1 EUR
ICNA0219052001 ICNA0219052010
Azure B (azure I), dark green powder, certified C.I. 52010
Azure B (Azure I)

Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.


Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.


Soluble in water (50 mg/ml) or ethanol (8-10 mg/ml); insoluble in diethyl ether.

Formula: C₁₅H₁₆ClN₃S
Melting Pt: 205…210 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011935
CAS Number: 531-55-5
EINECS: 208-511-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR