2-(4-Chlorophenoxy)isobutyric acid, white powder

Supplier: MP Biomedicals
Warning

Synonyms: 2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid

0215070180 0215070110
ICNA0215070180EA 102 EUR
ICNA0215070180 ICNA0215070110
2-(4-Chlorophenoxy)isobutyric acid, white powder
2-(4-Chlorophenoxy)isobutyric acid

Clofibric Acid reduces cholesterol levels in blood through diminishing the activity of NADPH.


Soluble in ethanol, ether, aceton, chloroform and methanol (50 mg/ml - clear, colourless solution).

Formula: C₁₀H₁₁ClO₃
MW: 214,65 g/mol
Melting Pt: 120…122 °C (lit.)
MDL Number: MFCD00004192
CAS Number: 882-09-7
EINECS: 212-925-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR