Karl Fischer Titration
Choice of compact volumetric or coulometric Karl Fisher titrators and related reagents for water determination.
- Volumetric Karl Fischer titrators are for medium to large volumes of water (mg range) combining fast and precise water content determination with easy operation
- Coulometric Karl Fischer titrators are for small water volumes (µg range) combine fast and precise water content determination with very simple operation
- Karl Fischer reagents
Product selection
Coulometric Karl Fischer Titrators
Routine Karl Fischer coulometric titrators combine fast and precise water content determination with very simple operation....
Karl Fischer Reagents
Karl Fischer application for water determination...